Learn More
Sino Biological Recombinant Canine IL-5 Protein (His Tag)
Supplier:  Sino Biological 70019D08B100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of canine IL5 (NP_001006951.1) (Met 1-Ser 134) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3325