Learn More
Sino Biological Recombinant Canine Her2/ERBB2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 70024D08H100
Encompass_Preferred
A DNA sequence encoding the canine ERBB2 (ABW99108.1) (Met1-Thr652) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3331