Learn More
Sino Biological Recombinant Canine EGFR Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 70026D08B50
Encompass_Preferred
A DNA sequence encoding the canine EGFR (XP_533073.3) (Met1-Ser618) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3334