Learn More
Sino Biological Recombinant Canine Angiopoietin-like 7 Protein (hFc Tag)
Supplier:  Sino Biological 70030D02H50
Encompass_Preferred
A DNA sequence encoding the canine ANGPTL7 (E2R2G1) (Met1-Pro344) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3340