Learn More
Sino Biological Recombinant Canine Angiopoietin-like 7 Protein (His Tag)
Supplier:  Sino Biological 70030D08H50
Encompass_Preferred
A DNA sequence encoding the canine ANGPTL7 (XP_852356.1) (Met1-Pro344) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3342