Learn More
Sino Biological Recombinant Canine ANGPTL1 Protein (hFc Tag)
Supplier:  Sino Biological 70032D01H50
Encompass_Preferred
A DNA sequence encoding the canine ANGPTL1 (E2RQY3) (Glu197-Asp491) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3344