Learn More
Sino Biological Recombinant Canine FGF1 Protein
Supplier:  Sino Biological 70039DNAE100
Encompass_Preferred
A DNA sequence encoding the canine FGF1 (XP_849274.1) (Phe16-Asp155) was expressed with a N-terminal Met.
Catalog No. 50-162-3353