Learn More
Sino Biological Recombinant Canine IGFBP5 Protein (hFc Tag)
Supplier:  Sino Biological 70042D02H50
Encompass_Preferred
A DNA sequence encoding the canine IGFBP5 (XP_038303831.1) (Met1-Glu271) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3356