Learn More
Sino Biological Recombinant Canine NOV/CCN3 Protein (His Tag)
Supplier:  Sino Biological 70043D08H100
Encompass_Preferred
A DNA sequence encoding the canine NOV(XP_532317.3) (Met1-Met353) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3357