Learn More
Sino Biological Recombinant Canine FGF-9 Protein (mFc Tag)
Supplier:  Sino Biological 70045D04H50
Encompass_Preferred
A DNA sequence encoding the canine FGF9 (NP_002001.1) (Leu4-Ser208) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-162-3360