Learn More
Sino Biological Recombinant Canine ACVR1 Protein (hFc Tag)
Supplier:  Sino Biological 70048D02H200
Encompass_Preferred
A DNA sequence encoding the canine ACVR1 (XP_549615.2) (Met1-Glu123) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3364