Learn More
Sino Biological Recombinant Canine ACVR1 Protein (His Tag)
Supplier:  Sino Biological 70048D08H200
Encompass_Preferred
A DNA sequence encoding the canine ACVR1 (XP_549615.2) (Met1-Glu123) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3366