Learn More
Sino Biological Recombinant Canine ALK-1 Protein (hFc Tag)
Supplier:  Sino Biological 70049D02H200
Encompass_Preferred
A DNA sequence encoding the canine ACVRL1(E2R174) (Met1-Gln119) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3368