Learn More
Sino Biological Recombinant Canine PDGF-A Protein (hFc Tag)
Supplier:  Sino Biological 70052D01H100
Encompass_Preferred
A DNA sequence encoding the canine PDGFA (NP_001177101.1) (Ser87-Arg196) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3371