Learn More
Sino Biological Recombinant Canine PDGF-B Protein (His Tag)
Supplier:  Sino Biological 70053D07Y50
Encompass_Preferred
A DNA sequence encoding the canine PDGFB (NP_001003383.1) (Ser82-Thr190) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3376