Learn More
Sino Biological Recombinant Canine CXCL16 Protein (His Tag)
Supplier:  Sino Biological 70060D08H50
Encompass_Preferred
A DNA sequence encoding the canine CXCL16(XP_849304.2) (Met1-Ser205) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3384