Learn More
Sino Biological Recombinant Canine CD40 Ligand/TNFSF5 Protein (mFc Tag)
Supplier:  Sino Biological 70068D04H100
Encompass_Preferred
A DNA sequence encoding the canine CD40LG (O97626) (Met112-Leu260) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-162-3389