Learn More
Sino Biological Recombinant Canine Fractalkine/CX3CL1 Protein (hFc Tag)
Supplier:  Sino Biological 70071D02H50
Encompass_Preferred
A DNA sequence encoding the canine CX3CL1 (H1ADY9) (Met1-Arg379) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3394