Learn More
Waters Corp Waters Vial, Polypropylene, 700 μL, Snap Neck, 8 x 40 mm
Supplier:  Waters Corp WAT022476
Encompass_Preferred
This polypropylene-molded vial with a 700 μL volume and a snap neck closure is supplied with a polyethylene septumless cap. It serves as a cost-effective alternative to glass vials and is ideal for applications where compounds might stick to glass surfaces.
  • Clean polypropylene-molded vial
  • Preferred in applications where compounds may stick to a polar glass surface
  • Cost-effective alternative to glass
  • Includes vial, cap, and septum
  • Autosampler vial
  • Total recovery vial
  • Snap neck closure
Catalog No. 50-785-156