Learn More
Waters Corp Waters Replacement Inserts, 96-well Plate, 700 μL, Glass
Catalog No. 50780697 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 700 μL glass inserts for 96-well plates solve limited sample needs. They replace original inserts, maximizing sample extraction by directing analytes for improved accessibility. Manufactured in an ISO-certified plant with ultra-pure reagents, batches are tested for reproducible performance. Inert glass prevents interference. They support diverse industries and HPLC platforms.
  • Addresses limited sample volume applications.
  • Replaces original glass insert of 96-well plates.
  • Maximizes extraction of required samples.
  • Suitable for pharmaceutical, environmental, biopharmaceutical, and other industries.
  • Manufactured in an ISO-certified plant using ultra-pure reagents.
  • Each batch is tested for excellent, reproducible performance.
  • Made from inert glass to prevent interference with analysis results.
Catalog No. 50-780-697 Supplier Waters Corp Supplier No. 186001437
These 700 μL glass inserts for 96-well plates solve limited sample needs. They replace original inserts, maximizing sample extraction by directing analytes for improved accessibility. Manufactured in an ISO-certified plant with ultra-pure reagents, batches are tested for reproducible performance. Inert glass prevents interference. They support diverse industries and HPLC platforms.
  • Addresses limited sample volume applications.
  • Replaces original glass insert of 96-well plates.
  • Maximizes extraction of required samples.
  • Suitable for pharmaceutical, environmental, biopharmaceutical, and other industries.
  • Manufactured in an ISO-certified plant using ultra-pure reagents.
  • Each batch is tested for excellent, reproducible performance.
  • Made from inert glass to prevent interference with analysis results.