Learn More
Medchemexpress LLC 7030B-C5 | 370871-42-4 | 99.8% | 359.38 g/mol | C17H21N5O4 | 10 MG
SDP
Catalog No. 5000288569
Encompass_Preferred
7030B-C5 is a small-molecule PCSK9 inhibitor intended for preclinical research. It shows in vitro activity (IC50 = 1.61 μM) and has been reported to reduce plasma cholesterol and triglyceride levels in vivo. The compound is identified by CAS No. 370871-42-4 and is supplied at high purity for laboratory studies.
  • Small-molecule PCSK9 inhibitor with reported IC50 of 1.61 μM.
  • Demonstrated in vivo activity reducing plasma cholesterol and triglyceride levels.
  • High purity (99.78%), suitable for preclinical studies.
  • Characterized molecular weight 359.38 g/mol and formula C17H21N5O4.
  • Supplied as a 10 mg quantity for laboratory use.
Catalog No. 50-002-88569 Supplier Medchemexpress LLC Supplier No. HY16193910MG
7030B-C5 is a small-molecule PCSK9 inhibitor intended for preclinical research. It shows in vitro activity (IC50 = 1.61 μM) and has been reported to reduce plasma cholesterol and triglyceride levels in vivo. The compound is identified by CAS No. 370871-42-4 and is supplied at high purity for laboratory studies.
  • Small-molecule PCSK9 inhibitor with reported IC50 of 1.61 μM.
  • Demonstrated in vivo activity reducing plasma cholesterol and triglyceride levels.
  • High purity (99.78%), suitable for preclinical studies.
  • Characterized molecular weight 359.38 g/mol and formula C17H21N5O4.
  • Supplied as a 10 mg quantity for laboratory use.