Learn More
Cole-Parmer Dow Corning 704 Vacuum Pump Oil, Silicone-Based; 500 mL
Supplier:  Cole-Parmer 7975410
Encompass_Preferred
Vacuum pump oil is formulated to ensure optimum pump performance and pump life. Dow Corning 704 is a single component oil for deep vacuum pumps (10-6 to 10-8 torr untrapped, and 10-10 to 10-11 torr trapped). Offers quick reaction to maximum vacuum levels, even after exposure to air within operating temperatures.
Catalog No. NC1535216