Learn More
Andwin Scientific 707 BLTPAD 7X10CM 50PK
SDP
Catalog No. NC9859509
Encompass_Preferred
The Grade 707 Blotting Pads are constructed of 100% cotton fiber. This product does not contain any additives and is cleaner than paper towels. It features a smooth, even texture that guarantees continuous contact of buffer from the chamber through t
Catalog No. NC9859509 Supplier Andwin Scientific Supplier No. 28298014
The Grade 707 Blotting Pads are constructed of 100% cotton fiber. This product does not contain any additives and is cleaner than paper towels. It features a smooth, even texture that guarantees continuous contact of buffer from the chamber through t