Learn More
MSC 70HDT 1 X30 TAPERED HI TMP SI
Catalog No. 501299631
Encompass
This 20 mil thick electrical tape features silicone rubber backing and a triangular, tapered edge profile. It is designed for high-temperature applications, withstanding up to 180 °C (356 °F), and provides optimized adherence for protecting high voltage cables from tracking.
  • RoHS 2011/65/EU compliant
  • Chemical and UV-resistant
  • Self-fusing, inorganic silicone adhesive for optimized adherence
  • Provides excellent resistance to arcs and flashes
  • Triangular, tapered edge profile with blue guideline for easy half-lapping applications
Catalog No. 50-129-9631 Supplier MSC Supplier No. 72361298
This 20 mil thick electrical tape features silicone rubber backing and a triangular, tapered edge profile. It is designed for high-temperature applications, withstanding up to 180 °C (356 °F), and provides optimized adherence for protecting high voltage cables from tracking.
  • RoHS 2011/65/EU compliant
  • Chemical and UV-resistant
  • Self-fusing, inorganic silicone adhesive for optimized adherence
  • Provides excellent resistance to arcs and flashes
  • Triangular, tapered edge profile with blue guideline for easy half-lapping applications