Learn More
Alkali Scientific 70L Automation Tips P70 for Ag
SDP
Supplier:  Alkali Scientific ABFT1070
Encompass_Preferred
Alkali Scientifichas designed its Agilent P70-compatible automation tips to maximize performance with Agilent Bravo and Agilent VPrep robotic liquid handlers These tips do not require a separate labware definition and VWorks automation control software protocols remain unchanged The 384-format tip is fully interchangeable with the original Agilent P70 automation pipette tip The longest of the 384-format tip types Alkali Scientifics Agilent P70-compatible tip is ideal for protocols requiring larger volumes per transfer Its extended tip length enables access to taller labware such as deep-well plates Excellent performance is achieved even with volumes as low as 1 L Alkali Scientificmanufactures its Agilent Bravo and Agilent VPrep-compatible P70 tips using the highest-grade virgin polypropylene Our advanced polishing techniques minimize liquid retention without using releasing agents or additives Additionally all Agilent Bravo and Agilent VPrep-compatible tip boxes feature conduc
Catalog No. 50-302-6551