Learn More
Ace Glass, Inc. 70mL porosity A extraction thimble
SDP
Catalog No. 50973434
Encompass_Preferred
70mL porosity A extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-973-434 Supplier Ace Glass, Inc. Supplier No. 681312
70mL porosity A extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.