Learn More
Ace Glass, Inc. 70mL porosity B extraction thimbles
SDP
Catalog No. 50973435
Encompass_Preferred
70mL porosity B extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-973-435 Supplier Ace Glass, Inc. Supplier No. 681314
70mL porosity B extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.