Learn More
Ace Glass, Inc. 70mL porosity C extraction thimble
SDP
Catalog No. 50973436
Encompass_Preferred
70mL porosity C extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-973-436 Supplier Ace Glass, Inc. Supplier No. 681316
70mL porosity C extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.