Learn More
Ace Glass, Inc. 70mL porosity D extraction thimble
SDP
Catalog No. 50978885
Encompass_Preferred
70mL porosity D extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-978-885 Supplier Ace Glass, Inc. Supplier No. 681318
70mL porosity D extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.