Learn More
Ace Glass, Inc. 70mL porosity E extraction thimble
SDP
Catalog No. 50973437
Encompass_Preferred
70mL porosity E extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-973-437 Supplier Ace Glass, Inc. Supplier No. 681320
70mL porosity E extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.