Learn More
Ace Glass, Inc. Acrylic safety shield and polyethylene safety cap for Michel-Miller chromatography columns. Cap is 70mm ID and fits#11 threaded columns. The shield is 45cm long.
SDP
Catalog No. 50978434
Encompass_Preferred
70mm I.D. x 45cm L Acrylic Safety Shield with polyethylene safety cap, for use with #11 threaded Michel-Miller chromatography columns. Safety shield insures safe operation at pressures to 300 psi. Shield wall thickness: 3mm. The top thread of the column is hung in a polyethylene cap; the cap is then held by the plastic shield. The shield should be secured with a clamp.
Catalog No. 50-978-434 Supplier Ace Glass, Inc. Supplier No. 5798240
70mm I.D. x 45cm L Acrylic Safety Shield with polyethylene safety cap, for use with #11 threaded Michel-Miller chromatography columns. Safety shield insures safe operation at pressures to 300 psi. Shield wall thickness: 3mm. The top thread of the column is hung in a polyethylene cap; the cap is then held by the plastic shield. The shield should be secured with a clamp.