Learn More
Biotage 70MM ALUM WEIGHING PANS 100/PK
SDP
Catalog No. NC9075568
Encompass
Aluminum weighing pans are ideal for evaporating your o&g extracts to dryness. unlike glass, aluminum does not absorb atmospheric moisture which can interfere with your gravimetric measurement and accuracy of your o&g analysis. aluminum weighing pans sizes: 70 mm pans for up to 55 ml of o&g sample extract.
Catalog No. NC9075568 Supplier Biotage Supplier No. 50002HT
Aluminum weighing pans are ideal for evaporating your o&g extracts to dryness. unlike glass, aluminum does not absorb atmospheric moisture which can interfere with your gravimetric measurement and accuracy of your o&g analysis. aluminum weighing pans sizes: 70 mm pans for up to 55 ml of o&g sample extract.