Learn More
Pipeline Packaging Corp 70MM HDPE SCREWCAP EPDM GASKET
SDP
Catalog No. NC3377355
Encompass_Preferred
Pipeline 70MM_NATURAL_HDPE_TAMPER EVIDENT SC76_SCREW CAP_EPDM GASKET GK-98W_@ 375_W/ 3/4" KNOCK OUT, 6TPI.Purchase in multiples of 48 or 64.
Catalog No. NC3377355 Supplier Pipeline Packaging Corp Supplier No. 99484
Pipeline 70MM_NATURAL_HDPE_TAMPER EVIDENT SC76_SCREW CAP_EPDM GASKET GK-98W_@ 375_W/ 3/4" KNOCK OUT, 6TPI.Purchase in multiples of 48 or 64.