Learn More
Chemglass Life Sciences Polyethylene Stopper for Dewar Flask, Fits CG-1593 & CG-1594-01, Codes -02, & -03
SDP
Catalog No. CG159803
Encompass_Preferred
Polyethylene Stopper for Dewar Flask, Fits CG-1593 & CG-1594-01, Codes -02, & -03. Vented polyethylene stopper to minimize evaporation from Dewar flasks.
Catalog No. CG159803 Supplier Chemglass Life Sciences Supplier No. CG159803
Polyethylene Stopper for Dewar Flask, Fits CG-1593 & CG-1594-01, Codes -02, & -03. Vented polyethylene stopper to minimize evaporation from Dewar flasks.