Learn More
CELLTREAT Scientific Products 70mm x 15mm Tissue Culture Treated Dish w/Grip Ring, Sterile
SDP
Catalog No. 50202034
Encompass_Preferred
CELLTREAT Scientific Products 70mm x 15mm Tissue Culture Treated Dish. Pronounced grip ring for improved handling. Cell Culture Dishes have thicker and more durable plastic than Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 7-8mL and growth area of 36.3cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 500/case.
Catalog No. 50-202-034 Supplier CELLTREAT Scientific Products Supplier No. 229670
CELLTREAT Scientific Products 70mm x 15mm Tissue Culture Treated Dish. Pronounced grip ring for improved handling. Cell Culture Dishes have thicker and more durable plastic than Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 7-8mL and growth area of 36.3cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 500/case.