Learn More
Nanocomposix BioPure Platinum Nanoparticles - Bare (Citrate) - 70 nm, 1 mg/mL in aq. 4 mM sodium citrate, 1 mL
SDP
Catalog No. NC2885952
Encompass_Preferred
As a global leader in platinum nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.70 nm platinum nanoparticles combine unique plasmonic optical properties with biocompatibility. These nanoparticles are useful in a broad range of applications from fuel cell technologies, therapies, catalysis, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. NC2885952 Supplier Nanocomposix Supplier No. PTCB701M
As a global leader in platinum nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.70 nm platinum nanoparticles combine unique plasmonic optical properties with biocompatibility. These nanoparticles are useful in a broad range of applications from fuel cell technologies, therapies, catalysis, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.