Learn More
Bellco Glass, Inc. 70uM Cell Strainer, universal, individually wrapped, sterile, 50/case
SDP
Catalog No. 502329767
Encompass_Preferred
70uM Cell Strainer, universal, individually wrapped, sterile, 50/case. Cell Strainers are ideal for obtaining uniform single cell suspension from various sources. Cell Strainers are made from nylon with 3 different mesh sizes (40µm, 70µm and 101µm), showing optimal performances in a variety of applications such as stem cell & primary cell preparation. • Ideal for stem cell and tissue-derived primary cell preparation• Fit into nearly all 50ml conical tubes including BD Falcon (tm), TPP and MidSci brands. • Tab molded within upper ring enables easy sterile handling• Packed individually• Sterilized• Non-Pyrogenic, DNase/RNase Free
Catalog No. 50-232-9767 Supplier Bellco Glass, Inc. Supplier No. 451400257
70uM Cell Strainer, universal, individually wrapped, sterile, 50/case. Cell Strainers are ideal for obtaining uniform single cell suspension from various sources. Cell Strainers are made from nylon with 3 different mesh sizes (40µm, 70µm and 101µm), showing optimal performances in a variety of applications such as stem cell & primary cell preparation. • Ideal for stem cell and tissue-derived primary cell preparation• Fit into nearly all 50ml conical tubes including BD Falcon (tm), TPP and MidSci brands. • Tab molded within upper ring enables easy sterile handling• Packed individually• Sterilized• Non-Pyrogenic, DNase/RNase Free