Learn More
Chemglass Life Sciences 70µm Cell Straining Kit, ReadyStrain™, w/ White Strainer, 50mL Tube and Screw Cap, Individually Wrapped, Sterile
SDP
Catalog No. 501979428
Encompass_Preferred
70m Cell Straining Kit, ReadyStrain; w/ White Strainer, 50mL Tube and Screw Cap; Individually Wrapped, Sterile. ReadyStrain provides all the necessary parts for filtration of cell suspensions and combes them into a single, preassembled, ready to use, sterile packet. These kits eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes strainers and caps). The individual sterile packets mean researchers are no longer forced to expose entire bags of 50mL tubes and corresponding caps in order to perform one filtration. Instead they can open a single ReadyStrain packet directly under the hood. Each kit contains a sterile SureStrain Strainer, pre-attached to a sterile 50mL tube, with a separate sterile screw-cap ready for assembly, processing and storage after the filtration is complete. Kits are supplied in easy open tear-strip bags and gamma sterilized.
Catalog No. 50-197-9428 Supplier Chemglass Life Sciences Supplier No. CLS2007K070
70m Cell Straining Kit, ReadyStrain; w/ White Strainer, 50mL Tube and Screw Cap; Individually Wrapped, Sterile. ReadyStrain provides all the necessary parts for filtration of cell suspensions and combes them into a single, preassembled, ready to use, sterile packet. These kits eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes strainers and caps). The individual sterile packets mean researchers are no longer forced to expose entire bags of 50mL tubes and corresponding caps in order to perform one filtration. Instead they can open a single ReadyStrain packet directly under the hood. Each kit contains a sterile SureStrain Strainer, pre-attached to a sterile 50mL tube, with a separate sterile screw-cap ready for assembly, processing and storage after the filtration is complete. Kits are supplied in easy open tear-strip bags and gamma sterilized.