Learn More
eMolecules​ 7143-01-3 | Methanesulfonic anhydride | Oakwood Chemicals | MFCD00007556 | 174.190 | C2H6O5S2 | 97.000 | CS(=O)(=O)OS(C)(=O)=O | 500g | 480138435
Catalog No. 5000161204 Shop All eMolecules Inc. Products
Encompass_Preferred
Methanesulfonic anhydride | Oakwood Chemicals | 7143-01-3 | MFCD00007556 | 174.190 | C2H6O5S2 | 97.000 | CS(=O)(=O)OS(C)(=O)=O | 500g | 480138435
Catalog No. 50-001-61204 Supplier eMolecules​ Supplier No. 480138435
Methanesulfonic anhydride | Oakwood Chemicals | 7143-01-3 | MFCD00007556 | 174.190 | C2H6O5S2 | 97.000 | CS(=O)(=O)OS(C)(=O)=O | 500g | 480138435