Learn More
Waters Corp Valve, High Pressure, Motorized, 3-Way, HPLC
Catalog No. 50818879 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The High Pressure Motorized Valve is a three-way valve with two common ports and one controllable port. It is designed for use in HPLC systems, specifically in the V1 (High Pressure Inject Valve) and V2 (High Pressure Syringe Valve) positions within the Fluids Pak. This valve is stepper motor driven.
  • Three-way valve design
  • Features two common ports
  • Includes one controllable port
  • Stepper motor driven for precise operation
  • Compatible with 717 PLUS Autosamplers
  • Compatible with 2695 and 2695D Separation Modules
  • Utilized as High Pressure Inject Valve and High Pressure Syringe Valve in Fluids Pak
Catalog No. 50-818-879 Supplier Waters Corp Supplier No. WAT045438
The High Pressure Motorized Valve is a three-way valve with two common ports and one controllable port. It is designed for use in HPLC systems, specifically in the V1 (High Pressure Inject Valve) and V2 (High Pressure Syringe Valve) positions within the Fluids Pak. This valve is stepper motor driven.
  • Three-way valve design
  • Features two common ports
  • Includes one controllable port
  • Stepper motor driven for precise operation
  • Compatible with 717 PLUS Autosamplers
  • Compatible with 2695 and 2695D Separation Modules
  • Utilized as High Pressure Inject Valve and High Pressure Syringe Valve in Fluids Pak