Learn More
Waters Corp Waters Performance Maintenance Kit, For 717 Autosampler, HPLC System
Catalog No. 50818993 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Performance Maintenance Kit for the 717 Autosampler contains all the Quality Parts necessary to keep your instrument operating at peak performance.
  • Contains all quality parts necessary to keep instrument operating at peak performance.
  • Includes one seal pack replacement kit.
  • Includes one syringe.
  • Includes one filter insert assembly.
  • Includes one 2.3 mm x 11.5 mm filter.
  • Includes two 2 mm x 7 mm filters.
Catalog No. 50-818-993 Supplier Waters Corp Supplier No. WAT052669
The Performance Maintenance Kit for the 717 Autosampler contains all the Quality Parts necessary to keep your instrument operating at peak performance.
  • Contains all quality parts necessary to keep instrument operating at peak performance.
  • Includes one seal pack replacement kit.
  • Includes one syringe.
  • Includes one filter insert assembly.
  • Includes one 2.3 mm x 11.5 mm filter.
  • Includes two 2 mm x 7 mm filters.