Learn More
Waters Corp Waters Corporation Sample Loop, 200 μL, Stainless Steel, for 717plus
Catalog No. 50819108 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 200μL sample loop is the standard sample loop that is installed in the 717 Plus Autosampler and it is also used in the LC Module 1.
  • Standard sample loop
  • Designed for 717 Plus Autosampler
  • Compatible with LC Module 1
  • Volume of 200 μL
  • Constructed from stainless steel
  • Suitable for HPLC systems
  • Recommended replacement as required
Catalog No. 50-819-108 Supplier Waters Corp Supplier No. WAT045584
The 200μL sample loop is the standard sample loop that is installed in the 717 Plus Autosampler and it is also used in the LC Module 1.
  • Standard sample loop
  • Designed for 717 Plus Autosampler
  • Compatible with LC Module 1
  • Volume of 200 μL
  • Constructed from stainless steel
  • Suitable for HPLC systems
  • Recommended replacement as required