Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 72-well Microtest (Terasaki) Plate

Catalog No. 07000402 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Count:
72
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Count
07-000-402 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-402 Supplier Greiner Bio-One Supplier No. 654180

missing translation for 'validMainframeMsg'

Suitable for all applications for serological determination of HLA antigens

Alphanumeric well coding
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape Flat
Capacity (Metric) 10 L
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment Tissue Culture
Type Microtest Plate
Well Count 72
For Use With (Application) HLA Antigen Determination
Includes Lid
No. per Case 270
No. per Pack 10
Temperature Range (Metric) -20°C to +60°C
Length (Metric) 83.3 mm
Width (Metric) 58 mm
Height (Metric) 11 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.