Learn More
Electron Microscopy Sciences MicroWell 72-Well Plate, Conical w/ Lid, 10 mcL, 0.013 cm2 Culture Area
SDP
Supplier:  Electron Microscopy Sciences 7043910
Encompass_Preferred
Terasaki format plates measuring 56x82 mm with 60 wells shaped like inverted truncated cones. Well surface is Nunclon treated for good cell attachment. Stackable tight-fitting lids.
Type: Conical w/ Lid
Volume: 10 µl
Culture Area: 0.013 cm2
Catalog No. 50-930-299