Learn More
Electron Microscopy Sciences MicroWell 72-Well Plate, Conical w/ Lid, 10 mcL, 0.013 cm2 Culture Area
SDP
Catalog No. 50930300
Encompass_Preferred
Terasaki format plates measuring 56x82 mm with 60 wells shaped like inverted truncated cones. Well surface is Nunclon treated for good cell attachment. Stackable tight-fitting lids.
Type: Conical w/ Lid
Volume: 10 µl
Culture Area: 0.013 cm2
Catalog No. 50-930-300 Supplier Electron Microscopy Sciences Supplier No. 7043915
Terasaki format plates measuring 56x82 mm with 60 wells shaped like inverted truncated cones. Well surface is Nunclon treated for good cell attachment. Stackable tight-fitting lids.
Type: Conical w/ Lid
Volume: 10 µl
Culture Area: 0.013 cm2