Learn More
Fire King Security Group Fire Resistant - Double door storage cabinet - 72"H Arctic White Finish
SDP
Catalog No. NC1533743
Encompass_Preferred
Double door storage cabinet. UL Class 350 fire rating and with one Medeco UL listed high-security keylock and equipped with 4 adjustable shelves. HIPAA compliance solution.
Catalog No. NC1533743 Supplier Fire King Security Group Supplier No. CF7236DAW
Add to Cart
Add to Cart
Request a Quote
Double door storage cabinet. UL Class 350 fire rating and with one Medeco UL listed high-security keylock and equipped with 4 adjustable shelves. HIPAA compliance solution.