Learn More
Chemglass Life Sciences Chemglass 72,000 mL Heavy-Wall 3-Neck Round Bottom Flask (CG-1522-78), 1-CN 55/50 outer, 2-SN 45/50 outer
SDP
Catalog No. 502733587
Encompass_Preferred
Chemglass CG-1522-78 is a heavy-wall 3-neck round bottom flask with a 72,000 mL capacity designed for large-scale laboratory reactions and distillations. The center neck accepts a CN 55/50 outer joint and the two side necks accept SN 45/50 outer joints, providing multiple attachment points for condensers, thermometers, and addition funnels.
  • 72,000 mL capacity for large-scale reactions and batch processing.
  • Heavy-wall round-bottom construction for enhanced strength and thermal performance.
  • Three necks allow simultaneous attachment of condensers, probes, and addition funnels.
  • Center neck with 55/50 outer joint and two side necks with 45/50 outer joints for standard tapered glassware connections.
  • Clear glass body for visual process monitoring.
  • Suitable for distillation, reflux, and large-scale synthesis assemblies.
Catalog No. 50-273-3587 Supplier Chemglass Life Sciences Supplier No. CG152278
Add to Cart
Add to Cart
Request a Quote
Chemglass CG-1522-78 is a heavy-wall 3-neck round bottom flask with a 72,000 mL capacity designed for large-scale laboratory reactions and distillations. The center neck accepts a CN 55/50 outer joint and the two side necks accept SN 45/50 outer joints, providing multiple attachment points for condensers, thermometers, and addition funnels.
  • 72,000 mL capacity for large-scale reactions and batch processing.
  • Heavy-wall round-bottom construction for enhanced strength and thermal performance.
  • Three necks allow simultaneous attachment of condensers, probes, and addition funnels.
  • Center neck with 55/50 outer joint and two side necks with 45/50 outer joints for standard tapered glassware connections.
  • Clear glass body for visual process monitoring.
  • Suitable for distillation, reflux, and large-scale synthesis assemblies.