Learn More
Chemglass Life Sciences 72000mL Glas-Col Heating Mantle, Series TM, Spherical, 2 @ 2000 Watts
SDP
Catalog No. CG1200015
Encompass_Preferred
72000mL Glas-Col Heating Mantle, Series TM, Spherical, 2 @ 2000 Watts. The Glas-Col Series TM spherical flask mantle covers only the flasks bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. The Series TM mantle is available with a bottom hole for flask drain on a special order basis. Terminal box models are available with switches and pilot lights. The 72 liter size mantles arae 230 Volt and are supplied complete with a 4 foot, 3 wire cord and locking connector. The 72 Liter sizes are furnished with a terminal box instead of a cord.
Catalog No. CG1200015 Supplier Chemglass Life Sciences Supplier No. CG1200015
72000mL Glas-Col Heating Mantle, Series TM, Spherical, 2 @ 2000 Watts. The Glas-Col Series TM spherical flask mantle covers only the flasks bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. The Series TM mantle is available with a bottom hole for flask drain on a special order basis. Terminal box models are available with switches and pilot lights. The 72 liter size mantles arae 230 Volt and are supplied complete with a 4 foot, 3 wire cord and locking connector. The 72 Liter sizes are furnished with a terminal box instead of a cord.