Learn More
Sarstedt Inc Screw cap micro tubes, 1.5 ml, PCR Performance Tested, Low protein-binding, 1000/case
Catalog No. NC1477442 Shop All Sarstedt Inc. Products
Encompass_Preferred
1.5mL Screw Cap Micro Tube Protein Low Binding with skirted base, knurls, assembled cap, printed writing space and graduation.
This item is not returnable. View return policy
Catalog No. NC1477442 Supplier Sarstedt Inc Supplier No. 72.703.600
1.5mL Screw Cap Micro Tube Protein Low Binding with skirted base, knurls, assembled cap, printed writing space and graduation.