Learn More
MSC Dow Corning 99179271 10.3 oz Cartridge Clear RTV Silicone Joint Sealant -76 to 356°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732
Catalog No. 501113812
Encompass
Dow Corning 99179271 10.3 oz Cartridge Clear RTV Silicone Joint Sealant -76 to 356°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732
Catalog No. 50-111-3812 Supplier MSC Supplier No. 00231647
Dow Corning 99179271 10.3 oz Cartridge Clear RTV Silicone Joint Sealant -76 to 356°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732